Would you like to log in again?
KIMBLE® Gas Washing Bottles, with Fritted Disc, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMBLE®
657750-2525 657750-5023 657750-1223 657750-2523 657750-5025
KT657750-5025EA 586.49 USD InStock
KT657750-5025 KT657750-2523 KT657750-2525 KT657750-1223 KT657750-5023
KIMBLE® Gas Washing Bottles, with Fritted Disc, DWK Life Sciences
Bottles Gas Washing Bottles
Tall form gas washing bottle with full length joint with hooks and fritted disc.
  • Available in two porosities: extra coarse (170-220 micron) and coarse (40-60 micron)
  • Supplied complete with hooks and two 2" springs
  • Manufactured from 33 expansion, low extractable borosilicate glass
Bottles contain a fritted disc for more uniform gas distribution throughout the absorbing material.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty