Would you like to log in again?
Black Phenolic Screw Caps, Kimble Chase
Supplier: Kimble Chase
75204G-24400 75206G-33400 75206G-20400 75206G-63400 75203G-28400 75203G-43400 75201G-89400 75201G-45400 75203G-33400 75201G-22400 75206G-89400 75205-38400 75205G-20400 75203G-24400 73809-15425 75204G-38400 75204G-33400 75202G-70400 75201G-53400 75201G-28400 75202G-58400 75204G-28400 75202G-33400 75204G-48400 75203G-63400 75205-28400 75206G-53400 75203G-20400 75203G-53400 75205G-24400 75202G-63400 75204G-70400 75206G-22400 75201G-48400 75203G-45400 75201G-33400 75205G-28400 75206G-48400 75202G-38400 75205-22400 75204G-58400 75202G-48400 73809-20400 75206G-24400 73809-13425 75205G-22400 75204G-45400 75201G-63400 75203G-89400 75202G-24400 75204G-22400 75206G-70400 75201G-58400 75206G-58400 73809-18400 75206G-28400 75204G-43400 75206G-45400 75201G-43400 75202G-89400 75203G-22400 73809-22400 75202G-28400 75206G-43400 75202G-43400 75203G-58400 75202G-45400 75205G-33400 75203G-48400 75205G-38400 75201G-20400 75204G-53400 75205-20400 75201G-70400 75205-24400 75201G-24400 75202G-53400 75203G-70400 75203G-38400 75202G-22400 75201G-38400 75204G-89400 75204G-63400 75204G-20400 75206G-38400 75202G-20400 75205-33400
89046-890CS 287.44 USD InStock
89046-890 89046-770 KT73809-22400 89046-892 89046-772 89046-894 89046-850 89046-774 89046-852 89046-896 89046-776 89046-898 89046-810 89046-854 89046-778 89046-812 89046-856 89046-814 89046-858 89046-804 89046-848 89046-806 89046-808 KT73809-15425 89046-780 89046-782 89046-860 89046-784 89046-862 89046-786 89046-820 89046-864 89046-788 89046-822 89046-866 89046-900 89046-868 89046-824 89046-902 89046-816 89046-818 89046-790 KT73809-20400 89046-792 89046-870 89046-794 KT73809-18400 89046-872 89046-796 89046-752 89046-830 89046-874 89046-754 89046-798 89046-876 89046-832 89046-756 89046-878 89046-834 89046-713 89046-758 89046-714 89046-836 89046-826 89046-828 KT73809-13425 89046-880 89046-760 89046-882 89046-762 89046-840 89046-884 89046-764 89046-720 89046-842 89046-886 89046-766 89046-722 89046-844 89046-800 89046-888 89046-768 89046-846 89046-802 89046-716 89046-838 89046-718
Black Phenolic Screw Caps, Kimble Chase
Closures Bottle Closures
PTFE/LDPE liners offer excellent solvent resistance
Pulp/vinyl liners are an economical source for general-purpose usage. Solid PE liners are ideal for strong acid or base samples. Cone-shaped taperseal liners provide excellent sealing capabilites in liquid applications. Tinfoil liners are best suited for environmental sampling applications. White rubber liners are resilient and moisture resistant, ideal for use in hermetic sealing. Temperature tolerant black phenolic screw caps are dimensionally stable and offer a wide array of chemical compatibilities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty