Would you like to log in again?
KIMBLE® KONTES® Cold Finger, Condenser, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
746065-0000
KT746065-0000EA 186.05 USD InStock
KT746065-0000
KIMBLE® KONTES® Cold Finger, Condenser, DWK Life Sciences
Condensers
Drip tip on the bottom directs drops into the collar of Hickman distillation heads.
  • Micro-adjustable
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
This cold finger condenser is used as an accessory for microscale Standard Taper 14/10 threaded kits.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty