Would you like to log in again?
KIMBLE® Conical Bottom Vials, DWK Lifesciences
Supplier: DWK Life Sciences
KIMBLE®
746105-0003 746105-0010
KT746105-0010CS 43.07 USD InStock
KT746105-0010 KT746105-0003
KIMBLE® Conical Bottom Vials, DWK Lifesciences
Vials
These conical bottom are ideal for use in sample collection areas.
  • Conical bottom with thin wall permits superb heat transfer
  • The vial has a threaded 14/10 outer joint
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with a blue, glass-filled nylon cap, a PTFE/silicone septum, and an ethylene propylene O-ring.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty