Would you like to log in again?
KIMBLE® MICROFLEX® Threaded Microscale Kits, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
747020-0000 747000-0000 747020-0005 747010-0000 747000-0005 747010-0005
KT747020-0005EA 818.56 USD InStock
KT747020-0005 KT747000-0000 KT747010-0005 KT747020-0000 KT747000-0005 KT747010-0000
KIMBLE® MICROFLEX® Threaded Microscale Kits, DWK Life Sciences
Microchemistry Kits
These organic chemistry kits feature threaded chaste connections that can be assembled 'rightside up' or 'upside down'.
Highly temperature-resistant, blue nylon connectors are ideal for use to 200 °C (392 °F).
Packaging: Kits are packed in a 34.3W×25.4D×6.4H cm (13½×10×2½") rugged polyethylene storage case with die-cut foam insert and easily fit in lab bench drawers.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty