Would you like to log in again?
KIMBLE® Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Supplier: DWK Life Sciences
747125-1324 747125-2014 747125-2024 747125-1319 747125-2019 747125-1314
KT747125-2019EA 58.09 USD InStock
KT747125-2019 KT747125-1319 KT747125-2014 KT747125-2024 KT747125-1314 KT747125-1324
KIMBLE® Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Adapters allow interfacing between outer Standard Taper jointed glassware with threaded microscale components.
  • Threaded on one end
  • With [ST] inner joint
  • Manufactured from borosilicate glass
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty