Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
KIMBLE® Threaded Compression Caps and Connectors, DWK Life Sciences
Supplier: DWK Life Sciences
747205-1520 747205-2020 747205-2024 747205-2022 747205-1315 747205-1320 747205-1313 747205-0808 747205-0813
KT747205-2024CS 38.08 USD InStock
KT747205-2024 KT747205-1320 KT747205-0808 KT747205-2022 KT747205-0813 KT747205-2020 KT747205-1315 KT747205-1313 KT747205-1520
KIMBLE® Threaded Compression Caps and Connectors, DWK Life Sciences
Fittings and Connectors
Open-top compression cap and connectors consist of glass-filled nylon.
  • Threaded
  • For use to 200°C (392°F)
  • PTFE seals
Connectors have PTFE seals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty