Would you like to log in again?
Accessories for KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Supplier: DWK Life Sciences
KONTES® MICROFLEX®
748012-0000 748009-1005 748012-1000 748007-1060 748008-0005 748003-1000 748005-1000 748010-0025 748009-0005 748008-1005 748001-0003 748013-0000 748004-0000 748013-1000 748014-1000 748006-1100 748005-1651 748005-0000 748010-1025 748005-1001 746001-0001 764100-1001 748001-0000 748002-0000 748030-0049 748017-0020 748025-1034 748033-0008 748020-1000 748003-0000 748004-1000 748001-1000 748006-0100 748014-0000
KT748009-1005CS 36.97 USD InStock
KT748009-1005 KT748014-1000 KT748006-1100 KT748003-1000 KT748010-0025 KT748004-0000 KT748001-0000 KT748020-1000 KT748005-1651 KT748002-0000 KT748001-0003 KT748013-0000 KT748008-0005 KT764100-1001 KT748005-0000 10024-800 KT748009-0005 KT748017-0020 KT748003-0000 KT748007-1060 KT748010-1025 KT748014-0000 KT748006-0100 KT748025-1034 KT748004-1000 KT748001-1000 KT748012-1000 KT748012-0000 KT748005-1000 KT748005-1001 KT746001-0001 KT748033-0008 KT748013-1000 KT748008-1005
Accessories for KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Microchemistry Kits
Test fabric is useful for testing dyes on a variety of fabrics
Three-foot length of fabric four inches wide features 8 strips of different fibers woven side by side. Filament acetate, bleached cotton, spun polyamide type 66, polyester disperse dyeable, spun polyacrylic, spun silk, spun viscose, and worsted wool.
Accessory for Microscale Williamson kit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty