Would you like to log in again?
Polyethylene Stoppers, Kimble Chase
Supplier: DWK Life Sciences
774240-0019 774240-0014 774240-0024 774240-0029
KT774240-0029PK 45.82 USD InStock
KT774240-0029 KT774240-0019 KT774240-0014 KT774240-0024
Polyethylene Stoppers, Kimble Chase
Stoppers PE/LDPE/HDPE Stoppers
Low cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers
Leak-proof, air-tight seals and wide top flange for ease of use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty