Would you like to log in again?
KIMBLE® Stirrer Bearings, PTFE, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
781010-2440 781019-2942
KT781019-2942EA 525.91 USD InStock
KT781019-2942 KT781010-2440
KIMBLE® Stirrer Bearings, PTFE, DWK Life Sciences
Stirrers Stirring Tools
These solid stirrer bearings are ideal for use with Kontes series KT786500-0261, KT786500-0262, KT786500-0263, KT786500-0264 glass 10 mm precision ground stirring shafts and 19 mm precision ground glass stirring shafts.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty