Would you like to log in again?
KIMBLE® Stirrer Bearings, PTFE and Glass, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
781050-2440 781050-3445
KT781050-2440EA 284.79 USD InStock
KT781050-2440 KT781050-3445
KIMBLE® Stirrer Bearings, PTFE and Glass, DWK Life Sciences
Stirrers Stirring Tools
These bearings are ideal for use with series KT786500-0261, KT786500-0262, KT786500-0263, KT786500-0264 glass 10 mm precision ground stirring shafts.
Threaded glass ground joint bearings seal against the stirrer shaft by means of a PTFE O-ring, a precision bore PTFE sleeve bearing and a PTFE tightening screw with lock nut and washer.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty