Would you like to log in again?
KIMBLE® Flex-Coupling Adapter for Glass Stirring Rods, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
788030-0000
KT788030-0000EA 233.57 USD InStock
KT788030-0000
KIMBLE® Flex-Coupling Adapter for Glass Stirring Rods, DWK Life Sciences
Stirrers Stirring Tools
This flex-coupling uses to glass stirring rods to be fix with the motor drive.
  • The glass shaft is secured by means of a silicone rubber insert
  • The top bushing will accept a motor rod and is tightened by means of an Allen screw
The chance of breakage will be reduced due to this coupling uses a 3" long epoxy-coated spring to compensate for misalignment and eliminates the need for special adapters.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty