Would you like to log in again?
KIMBLE® HI-VAC® Valves, with PTFE Plug, Kimble Chase
Supplier: DWK Life Sciences
KONTES® HI-VAC®
826510-0004 826510-0008 826510-0012 826500-0012 826500-0004 826310-0020 826500-0008
KT826510-0008EA 187.11 USD InStock
KT826510-0008 KT826500-0004 KT826500-0012 KT826510-0004 KT826510-0012 KT826500-0008 KT826310-0020
KIMBLE® HI-VAC® Valves, with PTFE Plug, Kimble Chase
Valves Vacuum Valves
For vacuum applications up to 5x10-7torr
The shaft is non-rotating and the primary seal is made with a Viton® O-ring on a tapered seat. PTFE shaft can be used continuously at system temperatures of 230°C (446°F) or intermittently at 275°C (527°F).
Designed with a precision external thread for fine control. Valve can be used as a bleed up to 1/2 turn. Full open is at 11/2 turns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty