Would you like to log in again?
KIMBLE® HI-VAC® Right-Angle Valve, with PTFE Plug and without Tip O-Ring, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
826610-0004 826610-0012 826610-0002 826610-0008
KT826610-0008EA 191.74 USD InStock
KT826610-0008 KT826610-0002 KT826610-0012 KT826610-0004
KIMBLE® HI-VAC® Right-Angle Valve, with PTFE Plug and without Tip O-Ring, DWK Life Sciences
Valves Vacuum Valves
This high-vacuum valve is for use in vacuum applications up to 5×10-7 Torr.
  • Similar to KT826510 series but without tip o-ring exposure
  • Designed with a precision external thread for fine control
  • Can be used as a bleed up to 1/2 turn
  • Full open position is at 11/2 turns
  • Primary seal is made with a FKM o-ring on a tapered seat
  • Non-rotating PTFE shaft can be used continuously at system temperatures of 230 °C or intermittently at 275 °C
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty