Would you like to log in again?
KONTES® Solid Pennyhead [ST] Stoppers, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KONTES®
850100-0022 850100-0016 850100-0027 850100-0008 850100-0019 850100-0013 850100-0038 850100-0009
KT850100-0016EA 31.41 USD InStock
KT850100-0016 KT850100-0027 KT850100-0038 KT850100-0013 KT850100-0009 KT850100-0008 KT850100-0019 KT850100-0022
KONTES® Solid Pennyhead [ST] Stoppers, Kimble Chase
Stoppers Glass Stoppers
Glass stoppers for use with flasks, mixing cylinders, separatory funnels, and more
Meet ASTM Specification E694.
All stoppers are solid.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty