Would you like to log in again?
Medium Length Hex Head Hollow Glass Stoppers, Kimble Chase
Supplier: DWK Life Sciences
850400-2926 850400-2425 850400-1420 850400-1922
KT850400-1922EA 103.99 USD InStock
KT850400-1922 KT850400-2425 KT850400-1420 KT850400-2926
Medium Length Hex Head Hollow Glass Stoppers, Kimble Chase
Stoppers Glass Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders, and separatory funnels
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Hollow, hexagonal head medium length Standard Taper stopper can be easily gripped, and may be set on the flattened top to avoid contamination of the ground surface.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty