Would you like to log in again?
KIMBLE® Safety Tip Needles with Blunt Ends, DWK Life Sciences
Supplier: DWK Life Sciences
868280-1801 868280-2201 868280-2001
KT868280-1801PK 7.58 USD InStock
KT868280-1801 KT868280-2201 KT868280-2001
KIMBLE® Safety Tip Needles with Blunt Ends, DWK Life Sciences
Syringe Needles
Blunt end safety tip needles are used for general purpose dispensing in laboratory applications.
  • Nonsterile stainless steel needle with an inert plastic luer hub
  • Blunt end for safety
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty