Would you like to log in again?
KIMBLE® DUALL® Tissue Grinders, with Glass Pestles and Plastic-Coated Glass Tubes, DWK Life Sciences
Supplier: DWK Life Sciences
KONTES® DUALL®
885460-0022 885460-0021 885460-0024 885460-0023 885460-0025 885460-0020
KT885460-0025CS 467.6 USD InStock
KT885460-0025 KT885460-0023 KT885460-0024 KT885460-0021 KT885460-0022 KT885460-0020
KIMBLE® DUALL® Tissue Grinders, with Glass Pestles and Plastic-Coated Glass Tubes, DWK Life Sciences
Homogenizers Tissue Grinders
Ideal for grinding connective tissue such as muscle, heart and lung.
  • The coating is transparent, allowing an unobstructed view of the homogenate
  • Clearance between pestles and tubes is 0.10 - 0.15 mm (0.004 to 0.006")
  • Replacement components are available and completely interchangeable
  • Manufactured from 33 expansion, low extractable borosilicate glass
These grinders combine both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate. The plastic safety coating on tube will contain the homogenate if the tube breaks or cracks during motor-driven homogenization.
Similar to 885450 series but with plastic safety coating.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty