Would you like to log in again?
Kimble® Potter-Elvehjem Tissue Grinder Components, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
885512-0021 885512-0020 885512-0022 885511-0021 885511-0023 885511-0020 885512-0023 885511-0022
KT885512-0021EA 96.47 USD InStock
KT885512-0021 KT885512-0022 KT885512-0020 KT885512-0023 KT885511-0023 KT885511-0022 KT885511-0021 KT885511-0020
Kimble® Potter-Elvehjem Tissue Grinder Components, DWK Life Sciences
Homogenizers Tissue Grinders
A graduated glass tube and a finely machined PTFE pestle comprise this popular style.
  • Graduations are fused into the glass
  • Replacement components are completely interchangeable
Radial serrations on the lower portion of the pestle deliver the homogenate into the cylindrical portion of the tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty