Would you like to log in again?
KONTES® Potter-Elvehjem Tissue Grinders with PTFE Pestles, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KONTES®
886000-0023 886000-0024 886000-0021 886000-0018 886000-0022 886000-0020 886000-0019
KT886000-0018EA 227.15 USD InStock
KT886000-0018 KT886000-0019 KT886000-0021 KT886000-0020 KT886000-0023 KT886000-0022 KT886000-0024
KONTES® Potter-Elvehjem Tissue Grinders with PTFE Pestles, Kimble Chase
Homogenizers Tissue Grinders
For homogenization of tissue samples
Sizes 21–24 have radial serrations on the lower portion of the pestle to deliver the homogenate into the cylindrical portion of the tube. Tubes and pestles are interchangeable, with a uniform 0.10–0.15mm clearance.
Grinders have a PTFE pestle and an unground borosilicate glass tube.
Ordering information: Special clearance pestles are available upon request; contact your sales representative for more information.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty