Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Derivatization Vials, DWK Life Sciences

Supplier: DWK Life Sciences
896820-0000
KT896820-0000EA 475.13 USD InStock
KT896820-0000
Derivatization Vials, DWK Life Sciences
Vials
Versatile reaction vial used for protein hydrolysis and derivatization.

  • PTFE cap with top port of 3 mm Ø
  • CTFE screw-type on-off valve
  • FKM O-ring
  • Temperature resistance upto 150 °C

Vials holds twelve 6×50 mm sample tubes.

Certifications: USP Type I and ASTM E438, Type I, Class A requirements.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty