Would you like to log in again?
KIMBLE® Extended Tip Pipettes, DWK Lifesciences
Supplier: DWK Life Sciences
KIMBLE®
897085-0007 897085-0009
KT897085-0007CS 155.8 USD InStock
KT897085-0007 KT897085-0009
KIMBLE® Extended Tip Pipettes, DWK Lifesciences
Pipettes Pasteur/Transfer Pipettes
Manufactured from 51 expansion borosilicate glass conforming to USP type I and ASTM E438, type I, class B requirements.
These extended tip NMR pipettes are for retrieving and adding samples in NMR tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty