Would you like to log in again?
KONTES® Safety-Coated Filtering Flasks, Kimble Chase
Supplier: DWK Life Sciences
KONTES®
953760-0122 953760-4002 953760-2002 953760-1002
KT953760-2002EA 376.5 USD InStock
KT953760-2002 KT953760-1002 KT953760-0122 KT953760-4002
KONTES® Safety-Coated Filtering Flasks, Kimble Chase
Flasks Filter Flasks
Flasks are manufactured from heavy-wall borosilicate glass and bonded to a plastic safety coating for extra safety during vacuum filtration
The safety coating is autoclavable to 121°C (275°F). All flasks have #2 side arm for connection to 6.4mm (1/4") I.D. flexible tubing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty