Would you like to log in again?
Microfiltration Funnel, Glass, Kimble Chase
Supplier: DWK Life Sciences
953781-0090 953751-0000
KT953751-0000EA 164.8 USD InStock
KT953751-0000 KT953781-0090
Microfiltration Funnel, Glass, Kimble Chase
Funnels Filtration Funnels Micro Filter Funnels
This 90mm assembly is designed for large sample volumes or samples with high particulate loads that would tend to clog a 47mm diameter filter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty