Would you like to log in again?
KIMBLE® ULTRA-WARE® 90 mm Microfiltration Assembly with Stainless Steel Support and Ground Joint, DWK Life Sciences
Supplier: DWK Life Sciences
ULTRA-WARE®
953855-1090
KT953855-1090EA 2675.96 USD InStock
KT953855-1090
KIMBLE® ULTRA-WARE® 90 mm Microfiltration Assembly with Stainless Steel Support and Ground Joint, DWK Life Sciences
Filters Membrane Filter Units
The 90 mm filtration apparatus with stainless steel support is recommended when filtering large volumes of viscous or proteinaceous solutions to give the maximum flow rate.
  • This unit is also used to produce ultra clean filtrate since the screen will not shed particles into the filtrate
  • Manufactured from 33 expansion, low extractable borosilicate glass
0.0055" opening size and 19.4% open area with 0.007" wire diameter.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty