Would you like to log in again?
ULTRA-WARE® Vacuum/Pressure Bottles, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE® ULTRA-WARE®
953900-0010
KT953900-0010EA 76.8 USD InStock
KT953900-0010
ULTRA-WARE® Vacuum/Pressure Bottles, KIMBLE®, DWK Life Sciences
Bottles Glass Bottles
This laboratory bottle has been specially designed for use under vacuum and at pressures up to 1.5 bar (at a maximum of 140 °C)
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
This vacuum- and pressure-resistant bottle is manufactured from borosilicate glass with a GL 45 screw thread.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty