Would you like to log in again?
KIMBLE® ULTRA-WARE® Economy Three-Hole Cap Systems, DWK Life Sciences
Supplier: DWK Life Sciences
953902-0502 953902-5002 953930-5002 953902-2003 953902-0252 953902-1002 953902-1003 953902-2002 953930-2002 953930-1002 953930-1003
KT953902-2003EA 2612.31 USD InStock
KT953902-2003 KT953902-1002 KT953902-2002 KT953902-1003 KT953902-0502 KT953930-5002 KT953930-1002 KT953930-1003 KT953902-5002 KT953902-0252 KT953930-2002
KIMBLE® ULTRA-WARE® Economy Three-Hole Cap Systems, DWK Life Sciences
Chromatography Systems Complete HPLC Systems
The ULTRA-WARE® Reservoir System with reservoir bottles are made up of Type I, Class A borosilicate glass and PTFE cap body.
The glass with a plastic safety coating blocks virtually all UV light up to 385 nm, preventing photodegradation of light sensitive mobile phases.
The connecting threads on the top of the cap use standard 1/4-28 flangeless fittings. Two of the ports have 1/8” through holes, the third port has a 1/16” through hole.
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Economy Three Hole Cap Systems are supplied with Economy Three Hole Cap, Ultra-Ware® flat bottom reservoir, All PEEK Inlet/Sparge filter, 2 µm, flangeless nut, ETFE, 1/4-28. 1/8”, flangeless ferrule, ETFE, 1/8”, nut plug, ETFE, 1/4-28, FEP Teflon® tubing, 1/8” O.D. x 1/16” I.D. x 10, TFE/propylene o-ring, polypropylene screw collar and asssembly instructions.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty