Would you like to log in again?
KIMBLE® ULTRA-WARE® Four Valve Filtration Delivery Caps, DWK Life Sciences
Supplier: DWK Life Sciences
ULTRA-WARE®
953981-0047
KT953981-0047EA 1361.85 USD InStock
KT953981-0047
KIMBLE® ULTRA-WARE® Four Valve Filtration Delivery Caps, DWK Life Sciences
Closures Bottle Closures
This cap combines the mobile phase filtration, sparging/degassing, storage and delivery functions.
  • All tubing connections are made in the back of the cap, reducing the typical clutter of tubing at the reservoirs
  • An integral check valve in the sparge port prevents the mobile phase from backing up into the gas lines
  • A pressure release valve prevents the reservoir from being accidentally over-pressurized
  • The 0.25"-28 fitting connections allow easy connection to any HPLC pump system
  • Filter diameter of 47 mm
Cap body is manufactured from glass-filled PTFE, cap will fit any reservoir or bottle with a GL 45 thread.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty