Would you like to log in again?
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Supplier: CPC (Colder Products Company)
MPC3301239 (25PK) MPC3301639 (25PK) SQCC221212M MPC3301239 (1/PK) MPX44024T39M MPC44024T39M 25PK MPC3301639 (1/PK) SQCX222424M MPX44012T39M 25PK MPX3301239 (25PK) MPX3301239 (1/PK) SQCX222416M MPX44024T39M (1/PK)
MFLX31604-27EA 37.5 USD InStock
MFLX31604-27 MFLX31604-26 MFLX00296-OOBLK MFLX00296-OPBLK MFLX00296-OQBLK MFLX31604-28 MFLX31604-30 MFLX31604-34 MFLX31604-26BLK MFLX31604-32 MFLX31604-27BLK MFLX31604-28BLK MFLX00296-OQ
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Fittings and Connectors
Attach directly to your sanitary hardware assemblies!
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • USP Class IV materials
  • Fully sterilizable
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. All models feature USP Class VI platinum-cured silicone gaskets.
These quick-disconnect couplings require a large fitting body and large insert (order separately).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty