Would you like to log in again?
Masterflex® Manifold Mixing Solenoid Valves, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
080T6-24-62-5 080T8-12-62 080T4-24-62 080T2-12-62 080T5-12-62 080T5-24-62 080T8-24-62-5 080T8-24-62 080T6-12-62 080T8-12-62-5 080T3-12-62-5 080T3-24-62-5 080T2-12-62-5 080T3-12-62 080T4-12-62 080T4-24-62-5 080T2-24-62 080T6-24-62 080T2-24-62-5 080T4-12-62-5
MFLX01356-12EA 973 USD InStock
MFLX01356-12 MFLX01356-23 MFLX01356-01 MFLX01356-02 MFLX01356-14 MFLX01356-03 MFLX01356-04 MFLX01356-16 MFLX01356-05 MFLX01356-07 MFLX01356-18 MFLX01356-19 MFLX01356-08 MFLX01356-09 MFLX01356-20 MFLX01356-10 MFLX01356-21 MFLX01356-00 MFLX01356-22 MFLX01356-11
Masterflex® Manifold Mixing Solenoid Valves, Avantor®
Valves Solenoid Valves
Stand up to aggressive fluids.
  • Excellent for multiple liquid or gas control systems with aggressive or high-purity fluids
  • PTFE or PEEK wetted parts
  • Continuous duty and high cycle life
  • Suitable for pressure or vacuum service
These normally closed solenoid mixing valves feature low power consumption, minimal dead volume, and fast response time for all your mixing needs. All valves come with 15" (26 gauge) PTFE coated lead wires. Special orders are available for valves with different diaphragm and body material, or larger flow values — call for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty