Would you like to log in again?
George Fischer Type 546 Ball Valves 42387
Supplier: GF Signet
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
161.546.343 175.546.334(TYPE-546) 175.546.332(TYPE-546) 161.546.341 161.546.344 161.546.342 163.546.351 163.546.353 163.546.352
MFLX01357-03EA 80.9 USD InStock
MFLX01357-03 MFLX01357-05 MFLX01371-08 MFLX01374-94 MFLX01357-07 MFLX01371-09 MFLX01374-92 MFLX01357-01 MFLX01371-10
George Fischer Type 546 Ball Valves 42387
Valves Ball Valves
Optimize safety with this true union ball valve.
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty