Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Plastic Ball Valves, Two-way Straight, PVC, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
PVC-638-4F4CJG-B PVC-638-4M4CJ-E PVC226-4SMC4SMC-F PVC 226-2F2F-B PVC-638-6MTB-B PVC-638-4F4M-E PVC-638-6CJG6CJG-E PVC-638-6MTB-E PVC-638-6B6B-B PVC-638-6CJ6CJ-E PVC-638-6F6CJG-B PVC-638-6M6CJG-E PVC-638-4F4CJ-B PVC226-6B6B-E PVC-638-4M4CJG-B PVC-638-6FTB-E PVC-638-4F4CJ-E PVC-638-6CJ6CJ-B PVC226-4B4B-F PVC-638-4FTB-E PVC226-4F4F-F PVC-638-6FTB-B PVC226-2F2F-F PVC-638-6F6M-B PVC-638-6F6F-B PVC 226-4SMC4SMC-B PVC 226-8B8B-B PVC226-2F2F-E PVC-638-4B4B-E PVC-638-6B6B-E PVC-638-4M4CJ-B PVC-638-TB4F-B PVC226-6B6B-F PVC-638-4CJ4CJ-E PVC-638-4MTB-E PVC226-4F4M-F PVC-638-6F6F-E PVC226-2F2M-F PVC 226-4SMC4SMC-E PVC-638-6M6F-E PVC-638-6F6CJG-E PVC-638-6CJG6CJG-B PVC-638-4F4CJG-E PVC 226-4B4B-B PVC-638-4B4B-B PVC-638-4CJ4CJ-B PVC226-8B8B-E PVC226-4F4M-E PVC-638-4MTB-B PVC-638-4F4F-B PVC226-4B4B-E PVC 226-8B8B-F PVC226-4F4F-E PVC 226-6CJG6CJG-B PVC226-2F2M-E PVC-638-4M4CJG-E PVC-638-4SMC4SMC-E PVC-638-4F4F-E PVC-638-6M6CJG-B
MFLX01379-10EA 18.9 USD InStock
MFLX01379-10 MFLX01377-30 MFLX01378-42 MFLX01379-12 MFLX01378-44 MFLX01377-32 MFLX01378-87 MFLX01378-40 MFLX01378-38 MFLX01377-26 MFLX01379-08 MFLX01378-78 MFLX01378-34 MFLX01378-36 MFLX01377-24 MFLX01378-30 MFLX01378-32 MFLX01378-28 MFLX01377-16 MFLX01377-18 MFLX01378-24 MFLX01378-26 MFLX01378-20 MFLX01377-96 MFLX01377-52 MFLX01377-98 MFLX01377-10 MFLX01378-22 MFLX01377-92 MFLX01377-94 MFLX01377-50 MFLX01377-90 MFLX01377-08 MFLX01378-16 MFLX01378-18 MFLX01377-01 MFLX01378-12 MFLX01377-88 MFLX01377-44 MFLX01377-03 MFLX01378-14 MFLX01377-40 MFLX01377-84 MFLX01377-42 MFLX01377-86 MFLX01378-10 MFLX01378-92 MFLX01377-80 MFLX01378-95 MFLX01377-82 MFLX01377-38 MFLX01378-06 MFLX01378-08 MFLX01379-14 MFLX01378-01 MFLX01378-89 MFLX01379-13 MFLX01378-04 MFLX01378-03
Masterflex® Plastic Ball Valves, Two-way Straight, PVC, Avantor®
Valves Ball Valves
Nonmetallic wetted parts ideal for water and mild aqueous solutions.

  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)

Ball valves are designed primarily for on/off service.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty