Would you like to log in again?
Parker Hannifin Ball Valves, Polypropylene
Supplier: Parker Hannifin
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PP4VFC2-MG PP4VFC4-MG PP4VFE4-MG PP4VFE6-MG PP6VTEU6-MG PP6VFC6-MG PP4VUC6-MG PP6VME6-MG PP4VFC6-MG PP6VUC6-MG PP6VFE4-MG PP6VME8-MG PP6VMC4-MG PP4VMC4-MG PP6VFE6-MG PP6VME4-MG PP4VUC4-MG PP4VMC6-MG PP4VTEU6-MG PP6VUC4-MG PP4VME4-MG PP4VME8-MG PP6VMC6-MG
MFLX01379-87EA 15.5 USD InStock
MFLX01379-87 MFLX01379-65 MFLX01379-53 MFLX01379-97 MFLX01379-75 MFLX01379-67 MFLX01379-45 MFLX01379-77 MFLX01379-55 MFLX01379-61 MFLX01379-71 MFLX01379-63 MFLX01379-41 MFLX01379-51 MFLX01379-73 MFLX01379-95 MFLX01379-81 MFLX01379-69 MFLX01379-47 MFLX01379-79 MFLX01379-57 MFLX01379-49 MFLX01379-59
Parker Hannifin Ball Valves, Polypropylene
Valves Ball Valves
Create a secure and easy push-to-connect connection.
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
These Parker Hannifin polypropylene (PP) ball valves feature an all plastic design is leak free and corrosion resistant.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty