Would you like to log in again?
Double-Open-End Silver/Carbon Filter Cartridges
Supplier: CALCO LTD
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S-3
MFLX01508-91EA 104 USD InStock
MFLX01508-91
Double-Open-End Silver/Carbon Filter Cartridges
Filters Filter Cartridges
Remove chlorine and organic chemicals for odor reduction and better taste.
  • Constructed with silver impregnated carbon to inhibit the growth of bacteria
Silver/Carbon Cartridge has silver impregnated in activated carbon to inhibit growth of bacteria. Removes solids, organic compounds, chlorine, tastes, and odors. For cold water only. Produces 3500 gallons prior to exhaustion. Max temperature is 77 °F (25 °C); max differential pressure is 10 psid.
Note: Cartridge does not work with DOE housing models 01508-40, -41, and -42.
* Psid refers to pounds per square inch pressure drop through the filter system.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty