Would you like to log in again?
Big Blue General-Purpose Pleated Filter Cartridges
Supplier: Big Blue
255494-43 155120-43 155405-43 155305-43
MFLX01508-86EA 38.4 USD InStock
MFLX01508-86 MFLX01512-88 MFLX01512-76 MFLX01512-74
Big Blue General-Purpose Pleated Filter Cartridges
Filters Filter Cartridges
Design offers higher flow rates and dirt-loading capacities than spun or wound depth cartridges, especially when removing lower micron-sized particles.
  • Remove non-gelatinous particles that fall into a narrow size range, typically in post filtration or polishing applications
Pleated cartridges separate viscous sediments and flat particulates. Use polyester cartridges for water filtration and polypropylene cartridges with harsher chemicals. Plastisol end caps.
Note: Cartridges are for sediment filtration only; they will not remove bacteria.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty