Would you like to log in again?
Masterflex® Direct Lift 2- and 3-way Solenoid Valves, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
01540-10 01540-08 01540-04 01540-01 01540-14 01540-17 01540-18 01540-12 01540-09 01540-11 01540-13 01540-02 01540-07
MFLX01540-18EA 478 USD InStock
MFLX01540-18 MFLX01540-07 MFLX01540-17 MFLX01540-09 MFLX01540-08 MFLX01540-10 MFLX01540-01 MFLX01540-12 MFLX01540-11 MFLX01540-14 MFLX01540-13 MFLX01540-02 MFLX01540-04
Masterflex® Direct Lift 2- and 3-way Solenoid Valves, Avantor®
Valves Solenoid Valves
Ideal for use with high-purity or corrosive fluids.
  • All wetted parts made of inert PTFE
  • Minimal dead volume reduces problems with trapped liquids
Rated for 26" Hg vacuum. Maximum fluid temperature is 158 °F (70 °C). All valves have 15" lead wires. Valves are compact and lightweight — mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty