Would you like to log in again?
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
2P04MT/1C 3P24MT/6C 6P04MT/6C 4P04MT/6C 4P04/6C 5P24/6C 2P04/6C 2P24/1C 5P04MT/1C 3P24MT/1C 6P04/1C 4P24/6C 3P04/1C 4P24MT/1C 3P04MT/1C 6P24/6C 5P24MT/6C 6P24MT/6C 4P04MT/1C 2P24/6C 4P04/1C 3P04/6C 6P24MT/1C 5P04MT/6C 2P04MT/6C 6P04/6C 2P24MT/1C 3P24/1C 5P24MT/1C 5P04/6C 4P24/1C 6P24/1C 6P04MT/1C 2P24MT/6C 4P24MT/6C 2P04/1C 3P04MT/6C
MFLX03214-76EA 407 USD InStock
MFLX03214-76 MFLX03214-10 MFLX03215-64 MFLX03214-52 MFLX03214-74 MFLX03214-30 MFLX03214-50 MFLX03215-80 MFLX03214-28 MFLX03215-18 MFLX03215-16 MFLX03214-46 MFLX03214-68 MFLX03215-36 MFLX03214-66 MFLX03215-12 MFLX03215-56 MFLX03215-34 MFLX03215-78 MFLX03214-64 MFLX03215-76 MFLX03215-10 MFLX03214-62 MFLX03215-74 MFLX03215-30 MFLX03215-52 MFLX03215-50 MFLX03214-80 MFLX03214-18 MFLX03215-28 MFLX03214-16 MFLX03214-36 MFLX03215-46 MFLX03215-68 MFLX03214-78 MFLX03214-12 MFLX03214-56
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Flow Meters
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty