Would you like to log in again?
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 150-mm, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
P14/1-N044-40G P04/1-N044-40G P24/1-N044-40C P14/1-N044-40ST P24/1-N044-40ST T54/1-N044-40S P14/1-N044-40C T54/1-044-40G P04/1-N044-40ST P24/1-N044-40G P04/1-N044-40C
MFLX03217-36EA 230 USD InStock
MFLX03217-36 MFLX03217-90 MFLX03294-36 MFLX03294-34 MFLX03294-32 MFLX03219-33 MFLX03217-32 MFLX03217-87 MFLX03219-35 MFLX03217-34 MFLX03219-37
Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 150-mm, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty