Would you like to log in again?
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
32462-55 32462-50 TPG TP2 0322630 03226-10 TP3
MFLX03226-30EA 73.5 USD InStock
MFLX03226-30 MFLX03226-10 MFLX03226-40 MFLX32462-50 MFLX03226-50 MFLX32462-55 MFLX03218-58
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Flow Meters
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty