Would you like to log in again?
Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
T51/1-N112-02G P21/1-N082-03G P01/1-N032-41G P11/1-N102-05ST P01/1-N102-05ST P01/1-N032-41ST P11/1-N042-15G P11/1-N062-01ST P11/1-N092-04G P11/1-N102-05G T51/1-N102-05S P21/1-N092-04G P21/1-N042-15G T51/1-N042-15S P21/1-N112-02G P21/1-N102-05ST P21/1-N042-15ST P21/1-N102-05G P11/1-N082-03G P01/1-N062-01G P11/1-N082-03ST P11/1-N112-02G T51/1-034-39G P01/1-N042-15G P11/1-N042-15ST P01/1-N102-05G P21/1-N112-02ST P21/1-N062-01ST P01/1-N034-39G P01/1-N034-39ST P11/1-N032-41ST P11/1-N034-39G P01/1-N092-04ST P21/1-N034-39ST P01/1-N082-03ST P11/1-N032-41G T51/1-092-04G T51/1-N082-03S T51/1-N034-39S P21/1-N032-41ST P21/1-N092-04ST P11/1-N092-04ST P21/1-N034-39G T51/1-082-03G P21/1-N032-41G P11/1-N062-01G P01/1-N082-03G T51/1-N032-41S P01/1-N112-02ST P01/1-N062-01ST P21/1-N062-01G T51/1-N032-41G P21/1-N082-03ST T51/1-N062-01S P01/1-N112-02G T51/1-N062-01G P11/1-N112-02ST T51/1-N112-02S T51/1-N092-04S P01/1-N092-04G P01/1-N042-15ST
MFLX03227-28EA 294 USD InStock
MFLX03227-28 MFLX03229-09 MFLX03227-26 MFLX03229-07 MFLX03227-24 MFLX03229-05 MFLX03229-13 MFLX03229-11 MFLX03227-30 MFLX03217-72 MFLX03217-73 MFLX03217-74 MFLX03217-76 MFLX03217-79 MFLX03295-28 MFLX03295-26 MFLX03295-24 MFLX03295-22 MFLX03295-20 MFLX03229-19 MFLX03229-17 MFLX03229-15 MFLX03229-25 MFLX03227-00 MFLX03229-23 MFLX03229-21 MFLX03227-84 MFLX03295-18 MFLX03295-16 MFLX03295-14 MFLX03295-12 MFLX03227-08 MFLX03295-10 MFLX03227-06 MFLX03227-04 MFLX03229-29 MFLX03229-27 MFLX03227-02 MFLX03227-12 MFLX03227-10 MFLX03229-31 MFLX03217-52 MFLX03217-53 MFLX03217-57 MFLX03295-08 MFLX03217-58 MFLX03295-06 MFLX03295-04 MFLX03295-02 MFLX03295-00 MFLX03227-18 MFLX03227-16 MFLX03227-14 MFLX03229-03 MFLX03227-22 MFLX03229-01 MFLX03227-20 MFLX03217-62 MFLX03217-63 MFLX03217-67 MFLX03217-68
Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty