Would you like to log in again?
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
P11/1-N044-40ST P01/1-N044-40C T51/1-N044-40S P01/1-N044-40G P21/1-N044-40ST T51/1-044-40G P11/1-N044-40G P21/1-N044-40G P01/1-N044-40ST P21/1-N044-4OC P11/1-N044-40C
MFLX03229-35EA 520 USD InStock
MFLX03229-35 MFLX03227-34 MFLX03227-32 MFLX03227-87 MFLX03229-33 MFLX03295-36 MFLX03295-34 MFLX03295-32 MFLX03217-78 MFLX03227-36 MFLX03229-37
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty