Would you like to log in again?
Masterflex® Correlated Variable Area Flowmeters, PTFE Housing, 65-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
T54/6-N014-96S T53/6-013-88G T54/6-013-88G T54/6-N013-88S T54/6-054-17G T54/6-N023-92S T51/6-N014-96G T51/6-N013-88G T51/6-N052-01G T51/6-N032-15S T53/6-052-01G T53/6-N032-15S T54/6-N042-07S T54/6-N022-13S T51/6-N013-88S T51/6-N012-10G T54/6-N052-01S T51/6-N042-07S T511/6-012-10S T54/6-032-15G T51/6-N054-17G T51/6-N022-13G T54/6-014-96G T54/6-012-10G T54/6-N032-15S T53/6-N013-88S T53/6-N012-10S T51/6-N064-63G T54/6-052-01G T54/6-N012-10S T51/6-N052-01S T51/6-N064-63S T53/6-014-96G T54/6-022-13G T51/6-N032-15G
MFLX03216-77EA 331 USD InStock
MFLX03216-77 MFLX32044-62 MFLX03216-55 MFLX03216-75 MFLX03216-52 MFLX03266-77 MFLX03266-55 MFLX32044-80 MFLX03216-50 MFLX32044-82 MFLX03216-70 MFLX03266-82 MFLX32044-77 MFLX32044-55 MFLX03266-80 MFLX32044-57 MFLX32044-50 MFLX03216-67 MFLX03216-87 MFLX03266-67 MFLX03216-65 MFLX32044-52 MFLX03216-85 MFLX03266-65 MFLX32044-90 MFLX03216-62 MFLX32044-70 MFLX03216-60 MFLX03216-82 MFLX03216-80 MFLX32044-65 MFLX32044-87 MFLX32044-88 MFLX32044-67 MFLX03216-57
Masterflex® Correlated Variable Area Flowmeters, PTFE Housing, 65-mm Scale, Avantor®
Flow Meters
Designed for high-purity applications with flexibility for a broad array of gases.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
The substitution of metal fittings with PTFE eliminates a potential contamination source for high-purity applications. The glass tube utilizes a fused ceramic scale for a precise, permanent measuring guide. In addition, a vertical-tangential locator line provides readings with hairline accuracy. Further improving readability is a front shield that magnifies the scale 16%. An anodized aluminum frame protects the heavy-walled glass flowtube; a white acrylic backplate protects and enhances viewing in the protective frame.
Select a flowmeter with a valve for flow rate control. The standard valve is suitable where high resolution metering is not essential. The high precision valve features a fine-adjust 16-turn “non-rising stem” to more accurately set your desired flow rate.
All flowmeters come with correlation data sheets for water and numerous gases (listed below) at standard temperature and pressure (STP).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty