Would you like to log in again?
Masterflex® Correlated Variable Area Flowmeters, 65-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
P13/6-N032-15G P03/6-N012-10G P03/6-N052-01G P23/6-N022-13ST P23/6-N032-15ST P13/6-N014-96G P23/6-N042-07G P03/6-N022-13G P03/6-N042-07ST P03/6-N014-96ST P03/6-N0365ST P03/6-N014-96G P13/6-N042-07G P03/6-N054-17ST P23/6-N032-15G P23/6-N012-10ST P03/6-N042-07G P23/6-N014-96G P03/6-N012-10ST P03/6-N013-88ST P13/6-N022-13ST P03/6-N023-92ST P23/6-N013-88G P23/6-052-01G P23/6-N022-13G P03/6-N0365G P13/6-N012-10ST P03/6-N023-92G P03/6-N013-88G P03/6-N032-15G P23/6-N0365ST P23/6-N012-10G P23/6-N023-92ST P23/6-N054-17ST P13/6-N023-92G P03/6-N032-15ST P23/6-N023-92G P13/6-N023-92ST P03/6-N022-13ST
MFLX03268-16EA 274 USD InStock
MFLX03268-16 MFLX03266-15 MFLX03268-17 MFLX03266-34 MFLX03268-15 MFLX03268-78 MFLX03268-35 MFLX03268-54 MFLX03266-30 MFLX03266-18 MFLX03266-19 MFLX03266-16 MFLX03268-18 MFLX03266-17 MFLX03268-19 MFLX03268-20 MFLX03268-64 MFLX03268-60 MFLX03268-05 MFLX03268-27 MFLX03266-26 MFLX03266-04 MFLX03266-02 MFLX03266-24 MFLX03268-01 MFLX03266-00 MFLX03266-22 MFLX03268-68 MFLX03268-21 MFLX03266-20 MFLX03268-66 MFLX03266-09 MFLX03268-09 MFLX03266-08 MFLX03268-07 MFLX03268-29 MFLX03266-28 MFLX03266-06 MFLX03268-50
Masterflex® Correlated Variable Area Flowmeters, 65-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty