Would you like to log in again?
Masterflex® Correlated Variable-Area Flowmeters, High-Flow, 150-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
T53/1-N044-40S P03/1-N044-40G P23/1-044-40C P23/1-N044-40ST P03/1-N044-40C P03/1-N044-40ST P13/1-N044-40ST P13/1-N044-40C T53/1-N044-40G
MFLX03269-37EA 348 USD InStock
MFLX03269-37 MFLX03267-36 MFLX03269-35 MFLX03267-34 MFLX03267-32 MFLX03267-87 MFLX03269-86 MFLX03269-84 MFLX03267-90
Masterflex® Correlated Variable-Area Flowmeters, High-Flow, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty