Would you like to log in again?
Masterflex® Correlated Variable-Area Flowmeters with Valve, 150-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
P04/1-N112-02G P04/1-N062-01G T54/1-102-05G P04/1-N032-41G T54/1-N062-01G P24/1-N034-39ST P04/1-N062-01ST P14/1-N112-02ST P24/1-N062-01G T54/1-082-03G T54/1-N092-04S PO4/1-N042-15ST T54/1-N112-02G T54/1-N032-41G P24/1-N032-41G P04/1-N092-04G P04/1-N092-04ST T54/1-N082-03S P14/1-N102-05G P14/1-N102-05ST T54/1-N062-01S P24/1-N042-15G P14/1-N082-03G P14/1-N062-01G T54/1-N032-41S P24/1-N082-03G P04/1-N082-03ST P14/1-N062-01ST P04/1-N112-02ST P24/1-N062-01ST PO4/1-N042-15G T54/1-092-04G P04/1-N034-39G P04/1-N082-03G P24/1-N032-41ST P04/1-N034-39ST T54/1-034-39G P24/1-N082-03ST P24/1-N112-02ST P24/1-N034-39G P24/1-N102-05ST P24/1-N112-02G T54/1-N042-15S P14/1-N032-41G T54/1-N112-02S P14/1-N034-39ST T54/1-N102-05S P14/1-N112-02G P24/1-N102-05G P24/1-N092-04ST P14/1-N032-41ST P24/1-N042-15ST P04/1-N102-05ST GF-8521-1606 T54/1-N034-39S P04/1-N102-05G P24/1-N092-04G P14/1-N042-15G P04/1-N032-41ST P14/1-N082-03ST
MFLX03219-05EA 360 USD InStock
MFLX03219-05 MFLX03217-26 MFLX03219-07 MFLX03294-26 MFLX03217-28 MFLX03219-09 MFLX03294-24 MFLX03217-70 MFLX03217-71 MFLX03217-30 MFLX03217-75 MFLX03219-11 MFLX03217-77 MFLX03219-13 MFLX03219-15 MFLX03294-18 MFLX03219-17 MFLX03294-16 MFLX03219-19 MFLX03294-14 MFLX03294-12 MFLX03294-10 MFLX03217-80 MFLX03217-81 MFLX03217-85 MFLX03219-21 MFLX03219-23 MFLX03217-00 MFLX03219-25 MFLX03217-02 MFLX03219-27 MFLX03217-04 MFLX03294-06 MFLX03219-29 MFLX03294-04 MFLX03217-06 MFLX03217-08 MFLX03294-00 MFLX03217-50 MFLX03219-31 MFLX03217-51 MFLX03217-10 MFLX03217-55 MFLX03217-12 MFLX03217-56 MFLX03294-08 MFLX03217-14 MFLX03217-16 MFLX03217-18 MFLX03294-30 MFLX03217-60 MFLX03217-61 MFLX03217-20 MFLX03219-01 MFLX03217-65 MFLX03217-66 MFLX03217-22 MFLX03219-03 MFLX03217-24
Masterflex® Correlated Variable-Area Flowmeters with Valve, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty