Would you like to log in again?
+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
Supplier: GF Signet
P51530-VO P51530-P0 P51530-P1 P51530-P2
MFLX05618-10EA 597 USD InStock
MFLX05618-10 MFLX05618-11 MFLX05618-13 MFLX32487-19
+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
Flow Meters
Open-core design ensures accurate, linear output to ±1%.
  • Eliminate cavitation and minimize pressure drop
Sensors contain an open-core rotor with a small magnet in each of the four blades. As they rotate, the magnets pass a coil in the sensor. The transducer generates a linear frequency output that is proportional to the flow velocity. This signal travels up to 200 feet without amplification.
Open-core design eliminates cavitation at flow velocities from 1 to 20 ft/second. Closed or solid paddle-wheel designs create air bubbles at these velocities, causing a nonlinear, nonrepeatable signal.
Sensor materials resist most chemicals. All models have PVDF rotor and VITON® O-ring; and include a 25-ft connecting cable.
ROTOR-X flow sensors are compatible with all of our line-, battery-, and sensor-powered flowmeters, accumulators, and controllers. Mount in any type of pipe using an installation fitting—order separately.
Delivery information: Includes 25-ft connecting cable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty