Would you like to log in again?
Foot-Operated Pump Dispensers for Barrels, Canisters, and Tanks
Supplier: BURKLE INC
5000-2200 5000-6000 5000-1500 5000-4016 5000-1800
MFLX06040-39EA 228 USD InStock
MFLX06040-39 MFLX06040-31 MFLX06040-43 MFLX06040-33 MFLX06040-35
Foot-Operated Pump Dispensers for Barrels, Canisters, and Tanks
Pumps Drum/Transfer Pumps
Keep hands-free for safe dispensing.
  • Large-volume foot-operated bellows for high pumping capacity
  • Universal bung constructed of soft PVC for 1-3/16" to 2-3/4" (30 to 70 mm) openings
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
Safely transfer liquids keeping both hands on the container you are filling. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty