Would you like to log in again?
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5005-1200 5005-7000 5005-4000 5005-1500 5005-3000 5005-1000 5005-6000 0600-3003
MFLX06040-49EA 221 USD InStock
MFLX06040-49 MFLX06040-29 MFLX06040-51 MFLX06040-52 MFLX06040-53 MFLX06040-45 MFLX06040-46 MFLX06040-47
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Pumps Drum/transfer Pumps
Dispense liquids safely and quickly from most storage containers.
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
These hand-operated pumps feature a universal soft PVC bung consisting of a conical center and two conical sleeves allowing for an airtight seal between the pump and most liquid storage container openings. The telescopic immersion tube adapts itself to the container height on insertion, ensuring good removal of residual liquid. Various tube diameters are available to meet your individual liquid transfer needs. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty