Would you like to log in again?
Bürkle Stainless Steel Barrel Pumps for Flammable Liquids
Supplier: BURKLE INC
5601-0801 5601-0400 5601-0800
MFLX06043-84EA 391 USD InStock
MFLX06043-84 MFLX06043-88 MFLX06043-89
Bürkle Stainless Steel Barrel Pumps for Flammable Liquids
Pumps Drum/Transfer Pumps
Safely transfer flammable and combustible liquids with ease.
  • Constructed of conductive 304 stainless steel for enhanced durability and safety
  • Ideal for filling combustible liquids in accordance with groups IIA and IIB
  • Maintenance-free hand-operated pump
Safely transfer flammable liquids from containers with a capacity of 60 to 1000 L into smaller containers. To enhance safety, these hand-operated pumps can be grounded during use to conduct electricity. Operating temperature range is 32 to 140 °F (0 to 60 °C).
There are two pump styles to choose from. Models with discharge tube only feature a spout-type outlet. Models with discharge tube and stopcock are suitable for fluid transfer in which the outlet container is further from the filling container. The stopcock also allows for on/off fluid control. The removable hose is 3.9 ft (1.2 m) in length with a NW 8 mm ball valve and 1/2” (12.8 mm) diameter discharge nozzle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty