Would you like to log in again?
Hand-Operated Chemical-Resistant Drum Pumps
Supplier: VWR International
5606-1001 5606-0601 5606-0600 5606-1000
MFLX06043-97EA 824 USD InStock
MFLX06043-97 MFLX06043-96 MFLX06043-99 MFLX06043-98
Hand-Operated Chemical-Resistant Drum Pumps
Pumps Drum/Transfer Pumps
Safely transfer aggressive chemicals.
  • PTFE and FEP wetted materials withstand harsh chemicals
  • Stopcock contains hazardous vapors and prevents dripping
These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty